2'-Hydroxyacetophenone ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
o-Hydroxyacetophenone, 2-Acetophenol, 2-Hydroxyphenyl Methyl Ketone, 1-(2-Hydroxyphenyl)ethan-1-one, o-Acetylphenol, 1-(2-Hydroxyphenyl)-1-oxoethane, 1-Ethanoyl-2-hydroxybenzol, 2-Acetylphenol
A12550.22
A12550.36
A12550.22EA
1100
CZK
A12550.22
A12550.36
2'-Hydroxyacetophenone ≥98%
2'-Hydroxyacetophenone
Purity: 98%
CAS No.: 118-93-4
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 106°(222°F)
Refractive Index: 1.5584
Melting Point: 3-6°
Boiling Point: 218°
Beilstein Reference: 386123
MDL No.: MFCD00002219
Vzorec:
C₈H₈O₂ Bod varu: 218 °C Bod tání: 3…6 °C Hustota: 1,130 Bod vzplanutí: 106 °C (222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002219 CAS číslo: 118-93-4 EINECS: 204-288-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...