3',5'-Dimethoxyacetophenone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-(3,5-Dimethoxyphenyl)ethan-1-one, 1-(3,5-Dimethoxyphenyl)ethanone
161721-1G
161721-5G
SIAL161721-1GEA
3370
CZK
SIAL161721-1G
SIAL161721-5G
3',5'-Dimethoxyacetophenone, Sigma-Aldrich®
3',5'-Dimethoxyacetophenone
Vzorec:
C₁₀H₁₂O₃ MW: 180,2 g/mol Bod varu: 290 °C (1013 hPa) Bod tání: 40…41 °C |
MDL Number:
MFCD00008739 CAS číslo: 39151-19-4 EINECS: 254-322-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...