Tisk…

3',5'-Dimethoxyacetophenone ≥97%

Dodavatel: Thermo Scientific

Synonyms: 1-(3,5-Dimethoxyphenyl)ethan-1-one, 1-(3,5-Dimethoxyphenyl)ethanone

B25224.03 B25224.06
B25224.03EA 2490 CZK
B25224.03 B25224.06
3',5'-Dimethoxyacetophenone ≥97%
3',5'-Dimethoxyacetophenone

Purity: 97% CAS No.: 39151-19-4 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 110°(230°F) Refractive Index: 1.5430 Melting Point: 39-41° Boiling Point: 290-291° Beilstein Reference: 2045586 MDL No.: MFCD00008739

Vzorec: C₁₀H₁₂O₃
MW: 180,2 g/mol
Bod varu: 290…291 °C
Bod tání: 39…41 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008739
CAS číslo: 39151-19-4
EINECS: 254-322-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR