3',4'-Dimethoxyacetophenone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone
M-Clarity™ quality level = MQ200
156639-100G
156639-25G
SIAL156639-100GEA
3280
CZK
SIAL156639-100G
SIAL156639-25G
3',4'-Dimethoxyacetophenone, Sigma-Aldrich®
3',4'-Dimethoxyacetophenone
Vzorec:
C₁₀H₁₂O₃ MW: 180.2 Bod varu: 286 Bod tání: 48…49 °C |
MDL Number:
MFCD00008737 CAS číslo: 1131-62-0 EINECS: 214-468-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...