4'-Ethoxyacetophenone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
275719-100G
275719-25G
SIAL275719-100GEA
13500
CZK
SIAL275719-100G
SIAL275719-25G
4'-Ethoxyacetophenone, Sigma-Aldrich®
4'-Ethoxyacetophenone
Vzorec:
C₁₀H₁₂O₂ Bod varu: 83 °C (0,04 mmHg) Bod tání: 39 °C |
MDL Number:
MFCD00009095 CAS číslo: 1676-63-7 EINECS: 216-825-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...