Tisk…

3'-Methoxyacetophenone ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: 3′-Methoxyacetophenone

A15604.14 A15604.22 A15604.36
A15604.14EA 817 CZK
A15604.14 A15604.22 A15604.36
3'-Methoxyacetophenone ≥97%
3'-Methoxyacetophenone

Purity: 97% CAS No.: 586-37-8 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 110°(230°F) Refractive Index: 1.5425 Melting Point: -7° Boiling Point: 239-241° Beilstein Reference: 878552 MDL No.: MFCD00008736

Vzorec: C₉H₁₀O₂
MW: 150.18
Bod varu: 239…241 °C
Bod tání: –7 °C
Hustota: 1.09
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008736
CAS číslo: 586-37-8
EINECS: 209-573-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR