Tisk…

2'-Methoxyacetophenone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

M9203-100G M9203-25G
SIALM9203-100GEA 5780 CZK
SIALM9203-100G SIALM9203-25G
2'-Methoxyacetophenone, Sigma-Aldrich®
2'-Methoxyacetophenone
Vzorec: C₉H₁₀O₂
Bod varu: 131 °C (1013 hPa)
Hustota: 1,09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008725
CAS číslo: 579-74-8
EINECS: 209-446-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR