Tisk…

2'-Nitroacetophenone ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone

A13208.09 A13208.18
A13208.09EA 1800 CZK
A13208.09 A13208.18
2'-Nitroacetophenone ≥97%
2'-Nitroacetophenone

Purity: 97% CAS No.: 577-59-3 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Flash Point: 110°(230°F) Refractive Index: 1.5515 Odour: Characteristic Melting Point: 25-28° Boiling Point: 158-160°/16mm Beilstein Reference: 1102322 MDL No.: MFCD00007144

Vzorec: C₈H₇NO₃
MW: 165,15 g/mol
Bod varu: 158…160 °C (16 mmHg)
Bod tání: 25…28 °C
Hustota: 1,24
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007144
CAS číslo: 577-59-3
EINECS: 209-414-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR