Benzoyl isothiocyanate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
261653-25G
261653-5G
SIAL261653-25GEA
6770
CZK
SIAL261653-25G
SIAL261653-5G
Benzoyl isothiocyanate, Sigma-Aldrich®
Benzoyl isothiocyanate
Vzorec:
C₈H₅NOS Bod varu: 252 °C (1013 hPa) Bod tání: –1 °C Hustota: 1,197 g/cm³ (16 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004815 CAS číslo: 532-55-8 EINECS: 208-540-0 UN: 2810 ADR: 6.1,III Merck Index: 13,01115 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...