Tisk…

4-Acetylbenzoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Acetylbenzoic Acid, 4'-Acetophenonecarboxylic acid

M-Clarity™ quality level = MQ100

177458-10G 177458-1G
SIAL177458-10GEA 6980 CZK
SIAL177458-10G SIAL177458-1G
4-Acetylbenzoic acid, Sigma-Aldrich®
4-Acetylbenzoic acid
Vzorec: C₉H₈O₃
MW: 164.16
Bod varu: 340
MDL Number: MFCD00002561
CAS číslo: 586-89-0
EINECS: 209-588-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR