Tisk…

4-Acetylbenzoic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Acetylbenzoic Acid, 4'-Acetophenonecarboxylic acid

A15239.03 A15239.06 A15239.14
A15239.03EA 1380 CZK
A15239.03 A15239.06 A15239.14
4-Acetylbenzoic acid ≥98%
4-Acetylbenzoic acid

Purity: 98+% CAS No.: 586-89-0 Molecular Formula: C9H8O3 Formula Weight: 164.16 Odour: Odourless Melting Point: 208-212° Beilstein Reference: 2207355 MDL No.: MFCD00002561

Vzorec: C₉H₈O₃
MW: 164.16
Bod varu: 340
Bod tání: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00002561
CAS číslo: 586-89-0
EINECS: 209-588-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR