Tisk…

4-Chlorobenzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (4-Chlorophenyl)methanol

M-Clarity™ quality level = MQ100

C27115-25G C27115-100G
SIALC27115-25GEA 2600 CZK
SIALC27115-25G SIALC27115-100G
4-Chlorobenzyl alcohol, Sigma-Aldrich®
4-Chlorobenzyl alcohol
Vzorec: C₇H₇ClO
MW: 142,58 g/mol
Bod varu: 234 °C (1013 hPa)
Bod tání: 67…70 °C
MDL Number: MFCD00004652
CAS číslo: 873-76-7
EINECS: 212-852-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR