Tisk…

4-Chlorobenzyl alcohol ≥99%

Dodavatel: Thermo Fisher Scientific

A11504.14 A11504.22
A11504.14EA 1640 CZK
A11504.14 A11504.22
4-Chlorobenzyl alcohol ≥99%
4-Chlorobenzyl alcohol

Purity: 99% CAS No.: 873-76-7 Molecular Formula: C7H7ClO Formula Weight: 142.59 Melting Point: 68-72° Boiling Point: 234° MDL No.: MFCD00004652

Vzorec: C₇H₇ClO
Bod varu: 234 °C
Bod tání: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00004652
CAS číslo: 873-76-7
EINECS: 212-852-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR