Tisk…

3,5-Dichlorobenzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

188735-5G 188735-25G
SIAL188735-5GEA 3250 CZK
SIAL188735-5G SIAL188735-25G
3,5-Dichlorobenzyl alcohol, Sigma-Aldrich®
3,5-Dichlorobenzyl alcohol
Vzorec: C₇H₆Cl₂O
MW: 177,03 g/mol
Bod varu: 264 °C (1013 hPa)
Bod tání: 82 °C
MDL Number: MFCD00004634
CAS číslo: 60211-57-6
EINECS: 262-109-1
UN: 2250
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR