Tisk…

3,5-Dichlorobenzyl alcohol ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A13527.06 A13527.14
A13527.06EA 1510 CZK
A13527.06 A13527.14
3,5-Dichlorobenzyl alcohol ≥98%
3,5-Dichlorobenzyl alcohol

Purity: 98% CAS No.: 60211-57-6 Molecular Formula: C7H6Cl2O Formula Weight: 177.03 Melting Point: 76-80° Beilstein Reference: 1936389

Vzorec: C₇H₆Cl₂O
Bod varu: 264 °C (1013 hPa)
Bod tání: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00004634
CAS číslo: 60211-57-6
EINECS: 262-109-1
UN: 2250
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR