2,4-Dichlorobenzyl alcohol ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
(2,4-Dichlorophenyl)methanol
A13338.14
A13338.22
A13338.36
A13338.14EA
1830
CZK
A13338.14
A13338.22
A13338.36
2,4-Dichlorobenzyl alcohol ≥99%
2,4-Dichlorobenzyl alcohol
Purity: 99%
CAS No.: 1777-82-8
Molecular Formula: C7H6Cl2O
Formula Weight: 177.03
Flash Point:
Vzorec:
C₇H₆Cl₂O MW: 177,03 g/mol Bod varu: 150 °C (25 mmHg) Bod tání: 57…61 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004606 CAS číslo: 1777-82-8 EINECS: 217-210-5 Merck Index: 14,3060 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...