Tisk…

2-Chloro-3-nitrotoluene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

C61984-1G C61984-5G
SIALC61984-1GEA 905 CZK
SIALC61984-1G SIALC61984-5G
2-Chloro-3-nitrotoluene, Sigma-Aldrich®
2-Chloro-3-nitrotoluene
Vzorec: C₇H₆ClNO₂
MW: 171,58 g/mol
MDL Number: MFCD00134653
CAS číslo: 3970-40-9
EINECS: 223-591-9
UN: 2433
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR