Tisk…

α-Chloro-3-nitrotoluene, Sigma-Aldrich®

Dodavatel: Merck

191167-10G 191167-50G
SIAL191167-10GEA 1330 CZK
SIAL191167-10G SIAL191167-50G
α-Chloro-3-nitrotoluene, Sigma-Aldrich®
α-Chloro-3-nitrotoluene
Vzorec: C₇H₆ClNO₂
MDL Number: MFCD00007272
CAS číslo: 619-23-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR