2-Chloro-p-toluidine ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-4-methylaniline, 4-Amino-3-chlorotoluene
B22935.06
B22935.14
B22935.06EA
1110
CZK
B22935.06
B22935.14
2-Chloro-p-toluidine ≥98%
2-Chloro-p-toluidine
Purity: 98%
CAS No.: 615-65-6
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point: 99°(210°F)
Refractive Index: 1.5750
Melting Point: 7°
Boiling Point: 223-225°
Beilstein Reference: 774514
UN No.: 3429
MDL No.: MFCD00007666
Vzorec:
C₇H₈ClN MW: 141,6 g/mol Bod varu: 223…225 °C Bod tání: 7 °C Hustota: 1,151 Bod vzplanutí: 99 °C (210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007666 CAS číslo: 615-65-6 EINECS: 210-440-7 UN: 3429 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...