3,5-Dichlorobenzylamine 94%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L12054.03
L12054.06
L12054.03EA
1260
CZK
L12054.03
L12054.06
3,5-Dichlorobenzylamine 94%
3,5-Dichlorobenzylamine
Purity: 97%
CAS No.: 39989-43-0
Molecular Formula: C7H7Cl2N
Formula Weight: 176.05
Refractive Index: 1.5760
Boiling Point: 141-144°/15mm
UN No.: 2735
MDL No.: MFCD00052681
Vzorec:
C₇H₇Cl₂N MW: 176,04 g/mol Bod varu: 141…144 °C (15 mmHg) Hustota: 1,320 Storage Temperature: Ambient |
MDL Number:
MFCD00052681 CAS číslo: 39989-43-0 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...