Tisk…

2,5-Dichlorobenzylamine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: (2,5-Dichlorophenyl)methanamine

M-Clarity™ quality level = MQ200

648892-1G 648892-5G
SIAL648892-1GEA 0 CZK
SIAL648892-1G SIAL648892-5G
2,5-Dichlorobenzylamine, Sigma-Aldrich®
2,5-Dichlorobenzylamine
Vzorec: C₇H₇Cl₂N
MW: 176,04 g/mol
Bod varu: 124…126 °C (13 mmHg)
Hustota: 1,2334 g/cm³ (20 °C)
MDL Number: MFCD00052391
CAS číslo: 10541-69-2
UN: 2922
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR