Tisk…

2,5-Dichlorobenzylamine 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (2,5-Dichlorophenyl)methanamine

L11972.03 L11972.06
L11972.03EA 1790 CZK
L11972.03 L11972.06
2,5-Dichlorobenzylamine 97%
2,5-Dichlorobenzylamine

Purity: 97% CAS No.: 10541-69-2 Molecular Formula: C7H7Cl2N Formula Weight: 176.05 Flash Point: 110°(230°F) Refractive Index: 1.5775 Odour: Amine-like Boiling Point: 74-76°/0.1mm Beilstein Reference: 2639302 UN No.: 2735 MDL No.: MFCD00052391

Vzorec: C₇H₇Cl₂N
MW: 176,04 g/mol
Bod varu: 74…76 °C (0,1 mmHg)
Hustota: 1,318
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052391
CAS číslo: 10541-69-2
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR