4-Chloro-2-(trifluoromethyl)aniline ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B21755.14
B21755.22
B21755.36
B21755.14EA
1280
CZK
B21755.14
B21755.22
B21755.36
4-Chloro-2-(trifluoromethyl)aniline ≥97%
4-Chloro-2-(trifluoromethyl)aniline
Purity: 97%
CAS No.: 445-03-4
Molecular Formula: C7H5ClF3N
Formula Weight: 195.57
Flash Point: 95°(203°F)
Refractive Index: 1.5070
Melting Point: 8-9°
Boiling Point: 66-67°/3mm
Beilstein Reference: 2366801
UN No.: 2810
MDL No.: MFCD00007841
Vzorec:
C₇H₅ClF₃N Bod varu: 66…67 °C (3 mmHg) Bod tání: 8…9 °C Hustota: 1,386 Bod vzplanutí: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007841 CAS číslo: 445-03-4 EINECS: 207-151-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...