Tisk…

4-Chloro-o-toluic acid ≥98%

Dodavatel: Thermo Scientific
Varování

H27262.03 H27262.06
H27262.03EA 920 CZK
H27262.03 H27262.06
4-Chloro-o-toluic acid ≥98%
4-Chloro-o-toluic acid

Purity: 98% CAS No.: 7499-07-2 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Melting Point: 170-174° MDL No.: MFCD00045852

Vzorec: C₈H₇ClO₂
MW: 170,6 g/mol
Bod tání: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00045852
CAS číslo: 7499-07-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR