6-Chloro-o-toluic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Chloro-6-methylbenzoic acid
L18348.06
L18348.03
L18348.06EA
8030
CZK
L18348.06
L18348.03
6-Chloro-o-toluic acid ≥97%
6-Chloro-o-toluic acid
Purity: 97%
CAS No.: 21327-86-6
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Melting Point: 103-105°
Beilstein Reference: 2614092
Vzorec:
C₈H₇ClO₂ MW: 170,6 g/mol Bod tání: 103…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045799 CAS číslo: 21327-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...