Tisk…

4'-Chloroacetophenone ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12115.30 A12115.0B
A12115.30EA 1710 CZK
A12115.30 A12115.0B
4'-Chloroacetophenone ≥98%
4'-Chloroacetophenone

Purity: 98+% CAS No.: 99-91-2 Molecular Formula: C8H7ClO Formula Weight: 154.60 Flash Point: 90°(194°F) Refractive Index: 1.5550 Melting Point: 17-20° Boiling Point: 230-232° Beilstein Reference: 386014 Merck Reference: 14,2116 UN No.: 3416 MDL No.: MFCD00000624

Vzorec: C₈H₇ClO
Bod varu: 230…232 °C
Bod tání: 17…20 °C
Hustota: 1,190
Bod vzplanutí: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000624
CAS číslo: 99-91-2
EINECS: 202-800-7
UN: 3416
ADR: 6.1,II
Merck Index: 14,2116

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR