Tisk…

ω,4-Dichloroacetophenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Chloro-1-(4-chlorophenyl)ethanone, α,4-Dichloroacetophenone, 2,4'-Dichloroacetophenone

M-Clarity™ quality level = MQ100

392006-100G 392006-25G
SIAL392006-100GEA 4000 CZK
SIAL392006-100G SIAL392006-25G
ω,4-Dichloroacetophenone, Sigma-Aldrich®
ω,4-Dichloroacetophenone
Vzorec: C₈H₆Cl₂O
MW: 189,04 g/mol
Bod varu: 270 °C (1013 hPa)
Bod tání: 98 °C
MDL Number: MFCD00018926
CAS číslo: 937-20-2
EINECS: 213-323-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR