3'-Chloroacetophenone ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A15592.06
A15592.14
A15592.22
A15592.06EA
1390
CZK
A15592.06
A15592.14
A15592.22
3'-Chloroacetophenone ≥98%
3'-Chloroacetophenone
Purity: 98+%
CAS No.: 99-02-5
Molecular Formula: C8H7ClO
Formula Weight: 154.60
Flash Point: 105°(221°F)
Refractive Index: 1.5506
Boiling Point: 227-229°
Beilstein Reference: 636318
UN No.: 3416
MDL No.: MFCD00000593
Vzorec:
C₈H₇ClO Bod varu: 227…229 °C Hustota: 1,191 Bod vzplanutí: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000593 CAS číslo: 99-02-5 EINECS: 202-721-8 UN: 3416 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...