2',4'-Dichloroacetophenone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2′,4′-Dichloroacetophenone
M-Clarity™ quality level = MQ100
178373-100G
178373-25G
SIAL178373-100GEA
4010
CZK
SIAL178373-100G
SIAL178373-25G
2',4'-Dichloroacetophenone, Sigma-Aldrich®
2',4'-Dichloroacetophenone
Vzorec:
C₈H₆Cl₂O Bod varu: 247 °C (1013 hPa) Bod tání: 42 °C Hustota: 1,32 g/cm³ (20 °C) |
MDL Number:
MFCD00000581 CAS číslo: 2234-16-4 EINECS: 218-780-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...