Tisk…

2',3'-Dichloroacetophenone ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-(2,3-Dichlorophenyl)ethanone

A12667.03 A12667.06
A12667.03EA 1280 CZK
A12667.03 A12667.06
2',3'-Dichloroacetophenone ≥98%
2',3'-Dichloroacetophenone

Purity: 98% CAS No.: 56041-57-7 Molecular Formula: C8H6Cl2O Formula Weight: 189.04 Refractive Index: 1.5600 Boiling Point: 106-108°/2mm MDL No.: MFCD00052988

Vzorec: C₈H₆Cl₂O
Bod varu: 106…108 °C (2 mmHg)
Hustota: 1,293
Storage Temperature: Ambient
MDL Number: MFCD00052988
CAS číslo: 56041-57-7
EINECS: 259-954-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR