Tisk…

ω-Chloroacetophenone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

77460-100G 77460-500G
SIAL77460-100GEA 1920 CZK
SIAL77460-100G SIAL77460-500G
ω-Chloroacetophenone, Sigma-Aldrich®
ω-Chloroacetophenone
Vzorec: (C₆H₅)C(O)CH₂Cl
Bod varu: 244 °C (1013 hPa)
Hustota: 1,324 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000933
CAS číslo: 532-27-4
EINECS: 208-531-1
UN: 1697
ADR: 6.1,II
Merck Index: 13,02132

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR