β-Chloropropiophenone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Chloropropiophenone
M-Clarity™ quality level = MQ200
335614-10G
335614-50G
SIAL335614-10GEA
1780
CZK
SIAL335614-10G
SIAL335614-50G
β-Chloropropiophenone, Sigma-Aldrich®
β-Chloropropiophenone
Vzorec:
(C₆H₅)C(O)CH₂CH₂Cl Bod varu: 263 °C (1013 hPa) Bod tání: 48…49 °C Hustota: 1,181 g/cm³ (20 °C) |
MDL Number:
MFCD00000990 CAS číslo: 936-59-4 EINECS: 213-317-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...