Tisk…

4-tert-Butylbenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-(tert-Butyl)benzaldehyde

M-Clarity™ quality level = MQ200

384038-25ML 384038-5ML
SIAL384038-25MLEA 2190 CZK
SIAL384038-25ML SIAL384038-5ML
4-tert-Butylbenzaldehyde, Sigma-Aldrich®
4-tert-Butylbenzaldehyde
Vzorec: C₁₁H₁₄O
MW: 162,23 g/mol
Bod varu: 58…60 °C (0,2 mmHg)
Hustota: 0,971 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00035742
CAS číslo: 939-97-9
EINECS: 213-367-9
UN: 3082
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR