Tisk…

3-Tolualdehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: m-Tolualdehyde, 3-Methylbenzaldehyde, m-Tolylaldehyde

M-Clarity™ quality level = MQ200

T35505-100G T35505-25G T35505-5G
SIALT35505-100GEA 7070 CZK
SIALT35505-100G SIALT35505-25G SIALT35505-5G
3-Tolualdehyde, Sigma-Aldrich®
3-Tolualdehyde
Vzorec: C₈H₈O
Bod varu: 199 °C (1013 hPa)
Hustota: 1,0535 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003374
CAS číslo: 620-23-5
EINECS: 210-632-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR