Tisk…

2-Bromobenzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2-Bromophenyl)methanol

M-Clarity™ quality level = MQ100

184276-10G 184276-50G
SIAL184276-10GEA 1850 CZK
SIAL184276-10G SIAL184276-50G
2-Bromobenzyl alcohol, Sigma-Aldrich®
2-Bromobenzyl alcohol
Vzorec: C₇H₇BrO
MW: 187,04 g/mol
Bod varu: 255 °C (1013 hPa)
Bod tání: 80…82 °C
MDL Number: MFCD00004600
CAS číslo: 18982-54-2
EINECS: 242-719-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR