Tisk…

4-Bromo-2-methylanisole ≥98%

Dodavatel: Thermo Fisher Scientific

B21621.09 B21621.18
B21621.09EA 1080 CZK
B21621.09 B21621.18
4-Bromo-2-methylanisole ≥98%
4-Bromo-2-methylanisole

Purity: 98+% CAS No.: 14804-31-0 Molecular Formula: C8H9BrO Formula Weight: 201.06 Melting Point: 66-69° Boiling Point: 124-125°/25mm MDL No.: MFCD01321139

Vzorec: C₈H₉BrO
Bod varu: 124…125 °C (25 mmHg)
Bod tání: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01321139
CAS číslo: 14804-31-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR