Tisk…

4-Bromo-3-methylanisole ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: 1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene

A12318.09 A12318.18 A12318.30
A12318.09EA 1580 CZK
A12318.09 A12318.18 A12318.30
4-Bromo-3-methylanisole ≥97%
4-Bromo-3-methylanisole

Purity: 97% CAS No.: 27060-75-9 Molecular Formula: C8H9BrO Formula Weight: 201.06 Flash Point: 110°(230°F) Refractive Index: 1.5610 Boiling Point: 111-112°/15mm Beilstein Reference: 1933865 MDL No.: MFCD00060687

Vzorec: C₈H₉BrO
MW: 201.06
Bod varu: 111…112 °C (15 mmHg)
Hustota: 1.450
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060687
CAS číslo: 27060-75-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR