Tisk…

α-Bromo-2-nitrotoluene, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: α-Bromo-o-nitrotoluene, 1-(Bromomethyl)-2-nitrobenzene, alpha-Bromo-2-nitrotoluene, 2-Nitrobenzyl bromide

M-Clarity™ quality level = MQ100

107794-25G 107794-5G
SIAL107794-25GEA 5810 CZK
SIAL107794-25G SIAL107794-5G
α-Bromo-2-nitrotoluene, Sigma-Aldrich®
α-Bromo-2-nitrotoluene
Vzorec: C₇H₆BrNO₂
MW: 216,03 g/mol
Bod tání: 45…47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007184
CAS číslo: 3958-60-9
EINECS: 223-558-9
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR