4-Bromo-o-toluidine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A15543.14
A15543.22
A15543.14EA
1680
CZK
A15543.14
A15543.22
4-Bromo-o-toluidine ≥98%
4-Bromo-o-toluidine
Purity: 98%
CAS No.: 583-75-5
Molecular Formula: C7H8BrN
Formula Weight: 186.06
Flash Point:
Vzorec:
C₇H₈BrN Bod varu: 240 °C Bod tání: 56…60 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007825 CAS číslo: 583-75-5 EINECS: 209-519-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...