Tisk…

2-Bromo-p-toluidine ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Bromo-4-methylaniline, 4-Amino-3-bromotoluene

A13818.09 A13818.18
A13818.09EA 1260 CZK
A13818.09 A13818.18
2-Bromo-p-toluidine ≥99%
2-Bromo-p-toluidine

Purity: 99% CAS No.: 583-68-6 Molecular Formula: C7H8BrN Formula Weight: 186.06 Flash Point: 110°(230°F) Refractive Index: 1.6030 Odour: Characteristic Melting Point: 14-16° Boiling Point: 240-242° Beilstein Reference: 1931711 UN No.: 2810 MDL No.: MFCD00007635

Vzorec: C₇H₈BrN
MW: 186,05 g/mol
Bod varu: 240…242 °C
Bod tání: 14…16 °C
Hustota: 1,494
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007635
CAS číslo: 583-68-6
EINECS: 209-515-7
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR