3-Bromobenzylamine 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
(3-Bromophenyl)methanamine
B24963.06
B24963.14
B24963.22
B24963.06EA
1630
CZK
B24963.06
B24963.14
B24963.22
3-Bromobenzylamine 95%
3-Bromobenzylamine
Purity: 97%
CAS No.: 10269-01-9
Molecular Formula: C7H8BrN
Formula Weight: 186.06
Refractive Index: 1.5885
Odour: Amine-like
UN No.: 2735
MDL No.: MFCD01026119
Vzorec:
C₇H₈BrN MW: 186,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01026119 CAS číslo: 10269-01-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...