2-Bromobenzylamine 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
(2-Bromophenyl)methanamine
A12412.14
A12412.06
A12412.14EA
8310
CZK
A12412.14
A12412.06
2-Bromobenzylamine 97%
2-Bromobenzylamine
Purity: 98%
CAS No.: 3959-05-5
Molecular Formula: C7H8BrN
Formula Weight: 186.06
Odour: Amine-like
Boiling Point: 118-119°/9mm
UN No.: 2735
MDL No.: MFCD00025572
Vzorec:
C₇H₈BrN MW: 186,05 g/mol Bod varu: 118…119 °C (9 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00025572 CAS číslo: 3959-05-5 UN: 2735 ADR: 8,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...