2-Bromobenzylamine 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(2-Bromophenyl)methanamine
350510050
350510250
ACRO350510050EA
2550
CZK
ACRO350510050
ACRO350510250
2-Bromobenzylamine 98%
2-Bromobenzylamine
Vzorec:
C₇H₈BrN MW: 186.05 Storage Temperature: Ambient |
MDL Number:
MFCD00025572 CAS číslo: 3959-05-5 UN: 2735 ADR: 8 III |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5880 to 1.5930 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...