Tisk…

α-Bromo-4-iodotoulene ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Iodobenzyl bromide, 1-(Bromomethyl)-4-iodobenzene, α-Bromo-p-iodotoulene

A18200.06 A18200.14
A18200.06EA 2670 CZK
A18200.06 A18200.14
α-Bromo-4-iodotoulene ≥97%
α-Bromo-4-iodotoulene

Purity: 97% CAS No.: 16004-15-2 Molecular Formula: C7H6BrI Formula Weight: 296.94 Odour: Irritating Melting Point: 78-82° Boiling Point: 140°/15mm Beilstein Reference: 2325160 UN No.: 3261 MDL No.: MFCD00209656

Vzorec: C₇H₆BrI
MW: 296,93 g/mol
Bod varu: 140 °C (15 mmHg)
Bod tání: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00209656
CAS číslo: 16004-15-2
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR