α-Bromo-3-iodotoulene, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
1-(Bromomethyl)-3-iodobenzene, α-Bromo-m-iodotoulene, 3-Iodobenzyl bromide
M-Clarity™ quality level = MQ100
427691-25G
427691-5G
SIAL427691-25GEA
11510
CZK
SIAL427691-25G
SIAL427691-5G
α-Bromo-3-iodotoulene, Sigma-Aldrich®
α-Bromo-3-iodotoulene
Vzorec:
C₇H₆BrI MW: 296,93 g/mol Bod tání: 50…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019016 CAS číslo: 49617-83-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...