α-Bromo-3-iodotoulene 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-(Bromomethyl)-3-iodobenzene, α-Bromo-m-iodotoulene, 3-Iodobenzyl bromide
A11140.06
A11140.14
A11140.22
A11140.06EA
3380
CZK
A11140.06
A11140.14
A11140.22
α-Bromo-3-iodotoulene 96%
α-Bromo-3-iodotoulene
Purity: 96%
CAS No.: 49617-83-6
Molecular Formula: C7H6BrI
Formula Weight: 296.94
Flash Point:
Vzorec:
C₇H₆BrI MW: 296,93 g/mol Bod varu: 106 °C (1 mmHg) Bod tání: 48…51 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019016 CAS číslo: 49617-83-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...