Tisk…

α-Bromo-4-(trifluoromethoxy)toluene ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1-(Bromomethyl)-4-(trifluoromethoxy)benzene, p-(Trifluoromethoxy)benzyl bromide, 4-(Trifluoromethoxy)benzyl bromide

A18335.06 A18335.14
A18335.06EA 3710 CZK
A18335.06 A18335.14
α-Bromo-4-(trifluoromethoxy)toluene ≥97%
α-Bromo-4-(trifluoromethoxy)toluene

Purity: 97% CAS No.: 50824-05-0 Molecular Formula: C8H6BrF3O Formula Weight: 255.04 Flash Point: 94°(201°F) Refractive Index: 1.4800 Melting Point: 22-24° Boiling Point: 63°/3mm Beilstein Reference: 2521451 UN No.: 3261 MDL No.: MFCD00061238

Vzorec: C₈H₆BrF₃O
Bod varu: 63 °C (3 mmHg)
Bod tání: 22…24 °C
Hustota: 1,594
Bod vzplanutí: 94 °C (201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061238
CAS číslo: 50824-05-0
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR