Tisk…

α-Bromo-3-(trifluoromethoxy)toluene ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: m-(Trifluoromethoxy)benzyl bromide, 1-(Bromomethyl)-3-(trifluoromethoxy)benzene, 3-(Trifluoromethoxy)benzyl bromide

B25061.03 B25061.06
B25061.03EA 872 CZK
B25061.03 B25061.06
α-Bromo-3-(trifluoromethoxy)toluene ≥98%
α-Bromo-3-(trifluoromethoxy)toluene

Purity: 98% CAS No.: 159689-88-0 Molecular Formula: C8H6BrF3O Formula Weight: 255.04 Flash Point: 88°(190°F) Refractive Index: 1.4780 Boiling Point: 178-180° Beilstein Reference: 8052203 UN No.: 3265 MDL No.: MFCD00061271

Vzorec: C₈H₆BrF₃O
MW: 255.03
Bod varu: 178…180 °C
Hustota: 1.572
Bod vzplanutí: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061271
CAS číslo: 159689-88-0
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR