4-Bromo-α,α,α-trifluorotoluene, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene
M-Clarity™ quality level = MQ100
152692-100G
152692-25G
152692-5G
SIAL152692-100GEA
3020
CZK
SIAL152692-100G
SIAL152692-25G
SIAL152692-5G
4-Bromo-α,α,α-trifluorotoluene, Sigma-Aldrich®
4-Bromo-α,α,α-trifluorotoluene
Vzorec:
C₇H₄BrF₃ MW: 225,01 g/mol Bod varu: 154 °C (1013 hPa) Hustota: 1,6347 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000398 CAS číslo: 402-43-7 EINECS: 206-943-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...