Tisk…

4-Bromo-3-fluorotoluene ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Bromo-2-fluoro-4-methylbenzene

A19142.09 A19142.14
A19142.09EA 2520 CZK
A19142.09 A19142.14
4-Bromo-3-fluorotoluene ≥98%
4-Bromo-3-fluorotoluene

Purity: 98% CAS No.: 452-74-4 Molecular Formula: C7H6BrF Formula Weight: 189.03 Flash Point: 74°(165°F) Refractive Index: 1.5310 Boiling Point: 187-188° Beilstein Reference: 2500154 MDL No.: MFCD00040940

Vzorec: C₇H₆BrF
MW: 189,03 g/mol
Bod varu: 187…188 °C
Hustota: 1,494
Bod vzplanutí: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040940
CAS číslo: 452-74-4
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR